Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PXT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152990.3 | NP_694535.2 | 5 | 849 | ||
XM_011514400.2 | XP_011512702.1 | 4 | 522 | |||
GenBank mRNA | AF486827.1 | - | 2 | 146 | ||
BC031105.1 | - | 5 | 850 | 5297409 | ||
BC107049.2 | - | 2 | 136 | 40003975 |