Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM26E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153711.3 | NP_714922.1 | 2 | 898 | ||
GenBank mRNA | AK094140.1 | - | 2 | 873 | ||
AK315572.1 | - | 2 | 870 | |||
BC032556.1 | - | 2 | 921 |