Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAJIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037225.2 | NP_001032302.1 | 7, 8 | 670 | ||
NM_001300803.1 | NP_001287732.1 | 4, 5 | 436 | |||
NM_001318808.1 | NP_001305737.1 | 7, 8 | 1384 | |||
XM_005273918.2 | XP_005273975.1 | 7, 8 | 664 | |||
XM_006718502.2 | XP_006718565.1 | 8 | 781 | |||
XM_006718503.3 | XP_006718566.1 | 8 | 667 | |||
XM_006718504.2 | XP_006718567.1 | 8 | 781 | |||
XM_006718505.2 | XP_006718568.1 | 5 | 547 | |||
XM_011544940.2 | XP_011543242.1 | 8 | 662 | |||
XM_011544941.2 | XP_011543243.1 | 1, 2 | 363 | |||
GenBank mRNA | AK093130.1 | - | 7, 8 | 1378 | ||
BC033501.1 | - | 4, 5 | 368 | |||
BC106951.2 | - | 6, 7 | 465 | 40035116 | ||
BC106952.1 | - | 6, 7 | 465 | 40035117 | ||
LC068587.1 | - | 6 | 508 |