Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010888.3 | NP_001010888.3 | 1 | 127 | ||
XM_011530939.2 | XP_011529241.1 | 2 | 372 | |||
XM_017029478.1 | XP_016884967.1 | 7 | 1638 | |||
XM_017029479.1 | XP_016884968.1 | 9 | 4931 | |||
XM_017029480.1 | XP_016884969.1 | 7 | 1672 | |||
XM_017029481.1 | XP_016884970.1 | 7 | 1667 | |||
XM_017029482.1 | XP_016884971.1 | 6 | 1577 | |||
XM_017029483.1 | XP_016884972.1 | 5 | 1373 | |||
XM_017029484.1 | XP_016884973.1 | 5 | 1005 | |||
GenBank mRNA | BC140767.1 | - | 1 | 127 | 9021635 | |
BC171781.1 | - | 1 | 127 | 9054435 | ||
BX647241.1 | - | 1 | 36 |