Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZKSCAN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012981.4 | NP_001012999.3 | 5 | 1831 | ||
XM_017023200.1 | XP_016878689.1 | 5 | 2251 | |||
GenBank mRNA | AK131342.1 | - | 5 | 1831 | ||
AK302399.1 | - | 5 | 1867 | |||
AK303472.1 | - | 5 | 2228 | |||
BC130007.1 | - | 5 | 1489 | 40132986 | ||
BC151139.1 | - | 5 | 1475 | 9057039 | ||
BX648785.1 | - | 5 | 1819 |