Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093730.1 | NP_001087199.1 | 11 | 1381 | ||
GenBank mRNA | DQ516347.1 | - | 11 | 1381 |