Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF587B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204818.1 | NP_001191747.1 | 3 | 449 | ||
GenBank mRNA | AK299091.1 | - | 3 | 430 | ||
U71363.1 | - | 4 | 443 |
Gene Symbol: ZNF552 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024762.3 | NP_079038.2 | 3 | 381 | ||
XM_005259267.3 | XP_005259324.1 | 3 | 559 | |||
GenBank mRNA | AK097041.1 | - | 3 | 381 | ||
AK293445.1 | - | 3 | 560 | |||
DA345819.1 | - | 3 | 381 |
Gene Symbol: ZNF814 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144989.1 | NP_001138461.1 | 3 | 486 | ||
XM_017027197.1 | XP_016882686.1 | 2 | 131 | |||
GenBank mRNA | AK300483.1 | - | 3 | 449 |