Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929698 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110619.1 | 2 | 865 | |||
GenBank mRNA | AK097865.1 | - | 2 | 865 |
Gene Symbol: NOL4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256798.1 | NP_001243727.1 | 4 | 856 | ||
XM_005260282.4 | XP_005260339.1 | 4 | 1142 | |||
XM_005260283.4 | XP_005260340.1 | 5 | 1166 | |||
XM_005260284.4 | XP_005260341.1 | 4 | 441 | |||
XM_005260285.4 | XP_005260342.2 | 4 | 1146 | |||
XM_005260286.4 | XP_005260343.1 | 4 | 1066 | |||
XM_006723702.3 | XP_006723765.1 | 5 | 533 | |||
XM_006723703.3 | XP_006723766.1 | 4 | 445 | |||
XM_011528562.2 | XP_011526864.1 | 4 | 850 | |||
XM_011528563.1 | XP_011526865.1 | 4 | 850 | |||
XM_017027669.1 | XP_016883158.1 | 4 | 436 | |||
GenBank mRNA | AK097804.1 | - | 6 | 1519 | ||
BQ067609.1 | - | 4 | 611 |