Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTATSF1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033884.1 | 1 | 1529 | |||
GenBank mRNA | AK128391.1 | - | 1 | 654 | ||
BX648511.1 | - | 1 | 649 |