Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015410.1 | 1, 2 | 204 | |||
GenBank mRNA | AK026189.1 | - | 1, 2 | 204 | ||
AK300802.1 | - | 1, 2 | 233 | |||
AK310212.1 | - | 2, 3 | 324 |