Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005159.4 | NP_005150.1 | 3 | 440 | ||
GenBank mRNA | AK056592.1 | - | 3 | 263 | ||
AK129892.1 | - | 3 | 263 | |||
AK129902.1 | - | 3 | 263 | |||
AK290616.1 | - | 3 | 263 | |||
BC009978.2 | - | 3 | 402 | 4121697 | ||
CR541795.1 | - | 2 | 197 | |||
DB504220.1 | - | 3 | 440 |
Gene Symbol: POTEE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083538.1 | NP_001077007.1 | 15 | 2343 | ||
XM_011511216.2 | XP_011509518.1 | 11 | 2067 | |||
XM_017004159.1 | XP_016859648.1 | 11 | 2098 | |||
XM_017004160.1 | XP_016859649.1 | 7 | 1066 | |||
XM_017004161.1 | XP_016859650.1 | 17 | 2780 |
Gene Symbol: POTEI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277406.1 | NP_001264335.1 | 15 | 2291 | ||
XM_011511670.2 | XP_011509972.1 | 15 | 2463 | |||
XM_017004732.1 | XP_016860221.1 | 16 | 2442 | |||
XM_017004733.1 | XP_016860222.1 | 10 | 1188 | |||
XM_017004734.1 | XP_016860223.1 | 8 | 1125 | |||
XM_017004735.1 | XP_016860224.1 | 3 | 85 |
Gene Symbol: POTEJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277083.1 | NP_001264012.1 | 15 | 2180 | ||
XM_011511683.2 | XP_011509985.1 | 15 | 2352 | |||
XM_017004741.1 | XP_016860230.1 | 16 | 2325 | |||
XM_017004742.1 | XP_016860231.1 | 10 | 1188 | |||
XM_017004743.1 | XP_016860232.1 | 8 | 1120 |
Gene Symbol: POTEM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145442.1 | NP_001138914.1 | 11 | 1972 | ||
GenBank mRNA | BC063888.1 | - | 3 | 632 |