Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142928.1 | NP_001136400.1 | 2 | 503 | ||
XM_006716095.2 | XP_006716158.1 | 2 | 723 | |||
GenBank mRNA | AK055954.1 | - | 2 | 419 | ||
AK098061.1 | - | 2 | 509 | |||
DB001467.1 | - | 2 | 513 |