Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUEDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271875.1 | NP_001258804.1 | 11 | 3096 | ||
NM_001292025.1 | NP_001278954.1 | 11 | 3401 | |||
XM_006721900.2 | XP_006721963.1 | 11 | 3711 | |||
XM_011524812.2 | XP_011523114.1 | 11 | 2484 | |||
XM_017024640.1 | XP_016880129.1 | 10 | 2982 | |||
XM_017024641.1 | XP_016880130.1 | 12 | 3113 | |||
GenBank mRNA | CR627470.1 | - | 5 | 2707 |