Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MALRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142308.2 | NP_001135780.2 | 40 | 6621 | ||
XM_011519453.2 | XP_011517755.1 | 41 | 7159 | |||
XM_011519454.1 | XP_011517756.1 | 41 | 6485 | |||
XM_011519455.2 | XP_011517757.1 | 40 | 6986 | |||
XM_017016182.1 | XP_016871671.1 | 40 | 6431 | |||
XM_017016184.1 | XP_016871673.1 | 29 | 4848 | |||
XM_017016185.1 | XP_016871674.1 | 29 | 4845 | |||
XM_017016186.1 | XP_016871675.1 | 22 | 3402 | |||
GenBank mRNA | AK297683.1 | - | 12 | 1504 | ||
BG192521.1 | - | 1 | 69 | |||
KC843478.1 | - | 40 | 6621 |