Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005843.5 | NP_005834.4 | 13 | 1684 | ||
GenBank mRNA | AF042273.1 | - | 13 | 1443 | ||
AK023490.1 | - | 1 | 906 | |||
AK292265.1 | - | 13 | 1379 | |||
AK292847.1 | - | 13 | 1421 | |||
AL133600.1 | - | 13 | 1446 | |||
BC028740.2 | - | 13 | 1684 | 5260998 |