Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNKSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297647.1 | NP_001284576.1 | 16, 17 | 1531 | ||
NM_001297648.1 | NP_001284577.1 | 16, 17 | 1505 | |||
NM_006314.2 | NP_006305.2 | 16, 17 | 1510 | |||
XM_011540484.2 | XP_011538786.1 | 16, 17 | 1490 | |||
XM_011540485.2 | XP_011538787.1 | 13, 14 | 1020 | |||
GenBank mRNA | AB209335.1 | - | 16, 17 | 1504 | ||
AF100153.1 | - | 16, 17 | 1488 | |||
AK001900.1 | - | 16, 17 | 1484 | |||
AK222817.1 | - | 16, 17 | 1489 | |||
AK222904.1 | - | 16, 17 | 1493 | |||
AK292646.1 | - | 16, 17 | 1492 | |||
BC011604.2 | - | 16, 17 | 1512 | 3139227 | ||
BC012797.2 | - | 16, 17 | 1512 | 3505293 | ||
BT006900.1 | - | 16, 17 | 1473 |