Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATG7 SIRNA Assays ›
Gene Symbol:
ATG7
Gene Name:
autophagy related 7
Gene Aliases:
APG7-LIKE, APG7L, GSA7
Chromosome Location:
Chr.3: 11272324 - 11564704 on Build GRCh38
UniGene ID:
Hs.38032
Species:
Human
siRNA ID
HSS173705
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATG7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136031.2
NP_001129503.2
12
1305
NM_001144912.1
NP_001138384.1
11
1188
NM_006395.2
NP_006386.1
12
1305
XM_006712931.3
XP_006712994.1
12
1248
XM_006712932.3
XP_006712995.1
13
1512
XM_006712933.3
XP_006712996.1
12
1485
XM_011533277.2
XP_011531579.1
14
1636
XM_011533278.2
XP_011531580.1
13
1341
XM_011533279.2
XP_011531581.1
14
1636
XM_011533280.2
XP_011531582.1
14
1636
XM_011533281.2
XP_011531583.1
14
1636
XM_011533282.2
XP_011531584.1
14
1636
XM_011533283.2
XP_011531585.1
14
1636
XM_011533284.2
XP_011531586.1
14
1636
XM_011533286.2
XP_011531588.1
14
1636
XM_017005542.1
XP_016861031.1
12
1222
XM_017005543.1
XP_016861032.1
12
1248
XM_017005544.1
XP_016861033.1
13
1510
XM_017005545.1
XP_016861034.1
12
1290
XM_017005546.1
XP_016861035.1
13
1341
XM_017005547.1
XP_016861036.1
14
1636
XM_017005548.1
XP_016861037.1
12
1248
XM_017005551.1
XP_016861040.1
12
1248
XM_017005553.1
XP_016861042.1
9
878
XM_017005554.1
XP_016861043.1
9
878
GenBank mRNA
AF094516.1
-
11
1213
AK075221.1
-
12
1232
AK298833.1
-
12
1183
AK303694.1
-
11
1115
AK314504.1
-
12
1222
AL122075.1
-
6
654
BC000091.2
-
12
1220
3504204
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein modification process
protein lipidation
C-terminal protein lipidation
cellular response to nitrogen starvation
cellular response to starvation
positive regulation of autophagy
protein transport
macroautophagy
positive regulation of macroautophagy
protein ubiquitination
positive regulation of protein modification process
piecemeal microautophagy of nucleus
suppression by virus of host autophagy
positive regulation of apoptotic process
late nucleophagy
positive regulation of protein catabolic process
defense response to virus
membrane fusion
cellular response to hyperoxia
regulation of mitophagy
ubiquitin activating enzyme activity
protein binding
transcription factor binding
Atg12 activating enzyme activity
Atg8 activating enzyme activity
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-qlhls:80/10.239.162.255:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0