Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303618.1 | NP_001290547.1 | 3 | 1031 | ||
NM_001303619.1 | NP_001290548.1 | 2 | 624 | |||
NM_006566.3 | NP_006557.2 | 4 | 1033 | |||
XM_005266642.3 | XP_005266699.1 | 4 | 1132 | |||
XM_005266643.3 | XP_005266700.1 | 2 | 226 | |||
XM_006722374.3 | XP_006722437.1 | 2 | 697 | |||
XM_017025524.1 | XP_016881013.1 | 4 | 7776 | |||
XM_017025525.1 | XP_016881014.1 | 2 | 508 | |||
XM_017025526.1 | XP_016881015.1 | 3 | 402 | |||
XM_017025527.1 | XP_016881016.1 | 2 | 1066 | |||
GenBank mRNA | AJ537553.1 | - | 3 | 613 | ||
AK313199.1 | - | 3 | 1032 | |||
BC074787.2 | - | 3 | 618 | 30915562 | ||
GU935335.1 | - | 4 | 780 | |||
GU935336.1 | - | 2 | 383 | |||
U56102.1 | - | 4 | 792 |