Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080866.2 | NP_543142.2 | 2 | 753 | ||
XM_017017159.1 | XP_016872648.1 | 2 | 1671 | |||
XM_017017160.1 | XP_016872649.1 | 2 | 485 | |||
GenBank mRNA | AB062418.1 | - | 2 | 586 | ||
AB074812.1 | - | 2 | 734 | |||
AF440402.1 | - | 2 | 721 | |||
AJ295270.1 | - | 2 | 545 | |||
AK074246.1 | - | 2 | 753 | |||
BC022379.1 | - | 2 | 753 | |||
BC126288.1 | - | 2 | 635 | 8992004 |