Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLITRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281503.1 | NP_001268432.1 | 2 | 1507 | ||
NM_052910.2 | NP_443142.1 | 1 | 1950 | |||
GenBank mRNA | AB067497.1 | - | 1 | 1949 | ||
AK299479.1 | - | 1 | 1091 | |||
AY358289.1 | - | 1 | 1230 | |||
BC051738.1 | - | 1 | 1851 | 4816570 |