Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282175.1 | NP_001269104.1 | 10 | 1235 | ||
NM_001282176.1 | NP_001269105.1 | 10 | 1259 | |||
NM_001294.3 | NP_001285.1 | 10 | 1386 | |||
GenBank mRNA | AF037339.1 | - | 10 | 1229 | ||
AK074935.1 | - | 10 | 1230 | |||
AK075262.1 | - | 10 | 1229 | |||
AK223553.1 | - | 10 | 1230 | |||
AK293314.1 | - | 10 | 1251 | |||
AK304480.1 | - | 10 | 1218 | |||
AK316094.1 | - | 10 | 1255 | |||
BC004865.2 | - | 10 | 1265 | 3840991 | ||
BC012359.1 | - | 10 | 1219 | 4685026 | ||
BT007262.1 | - | 10 | 1214 | |||
KU177986.1 | - | 10 | 1214 | |||
KU177987.1 | - | 10 | 1214 |