Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242929.1 | NP_001229858.1 | 3 | 451 | ||
NM_001308089.1 | NP_001295018.1 | 5 | 641 | |||
NM_001324129.1 | NP_001311058.1 | 2 | 279 | |||
NM_001324130.1 | NP_001311059.1 | 5 | 849 | |||
NM_133450.3 | NP_597707.1 | 6 | 843 | |||
NR_040252.1 | 5 | 846 | ||||
XM_005255119.3 | XP_005255176.1 | 4 | 543 | |||
XM_011522372.1 | XP_011520674.1 | 5 | 773 | |||
XM_011522373.1 | XP_011520675.1 | 5 | 655 | |||
XM_011522374.1 | XP_011520676.1 | 4 | 389 | |||
XM_011522378.1 | XP_011520680.1 | 4 | 400 | |||
XM_017022935.1 | XP_016878424.1 | 3 | 451 | |||
XM_017022936.1 | XP_016878425.1 | 4 | 653 | |||
GenBank mRNA | AB075857.1 | - | 3 | 1279 | ||
AK057329.1 | - | 3 | 444 | |||
AK123496.1 | - | 6 | 844 | |||
AK301683.1 | - | 4 | 471 | |||
BC050387.1 | - | 5 | 482 | 5751728 | ||
BM545219.1 | - | 5 | 692 | |||
BX333650.2 | - | 3 | 296 | |||
DR004376.1 | - | 4 | 579 |