Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083614.1 | NP_001077083.1 | 5, 6 | 1097 | ||
NM_001308211.1 | NP_001295140.1 | 5, 6 | 1097 | |||
NR_003501.1 | 5, 6 | 1097 | ||||
XM_011545738.1 | XP_011544040.1 | 5, 6 | 1387 | |||
XM_011545739.1 | XP_011544041.1 | 5, 6 | 1006 | |||
GenBank mRNA | AB075850.1 | - | 5, 6 | 1054 | ||
AK095998.1 | - | 5, 6 | 1086 | |||
AL832489.1 | - | 5, 6 | 1073 | |||
BC040013.1 | - | 5, 6 | 1066 | 5728326 | ||
BX648129.1 | - | 3, 4 | 776 |