Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22orf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173793.4 | NP_776154.3 | 3 | 832 | ||
GenBank mRNA | AK095271.1 | - | 3 | 292 | ||
AK290073.1 | - | 3 | 805 | |||
BC062599.1 | - | 3 | 297 | 5248209 | ||
BX337505.2 | - | 3 | 831 | |||
CR456339.1 | - | 3 | 293 |