Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153605.3 | NP_705833.3 | 16 | 8199 | ||
XM_005247117.4 | XP_005247174.1 | 15 | 7902 | |||
GenBank mRNA | AF451988.1 | - | 4 | 328 | ||
AK091199.1 | - | 8 | 670 | |||
AL831927.1 | - | 4 | 222 | |||
BX647079.1 | - | 8 | 687 | |||
BX648353.1 | - | 13 | 2249 | |||
CR749313.1 | - | 13 | 1324 |