Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHODL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204174.1 | NP_001191103.1 | 4 | 632 | ||
NM_001204175.1 | NP_001191104.1 | 4 | 598 | |||
NM_001204176.1 | NP_001191105.1 | 5 | 698 | |||
NM_001204177.1 | NP_001191106.1 | 4 | 598 | |||
NM_001204178.1 | NP_001191107.1 | 5 | 698 | |||
NM_024944.2 | NP_079220.2 | 4 | 1007 | |||
XM_011529453.2 | XP_011527755.1 | 4 | 1241 | |||
XM_011529457.2 | XP_011527759.1 | 5 | 1004 | |||
XM_017028273.1 | XP_016883762.1 | 4 | 627 | |||
GenBank mRNA | AF257472.1 | - | 4 | 1007 | ||
AF523313.1 | - | 5 | 697 | |||
AF523314.1 | - | 5 | 698 | |||
AF523315.1 | - | 4 | 599 | |||
AK022689.1 | - | 5 | 880 | |||
AK296009.1 | - | 4 | 632 | |||
AK313725.1 | - | 4 | 655 | |||
AY358608.1 | - | 4 | 993 | |||
BC009418.1 | - | 4 | 977 | 3509743 |