Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR51E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152430.3 | NP_689643.2 | 2 | 472 | ||
GenBank mRNA | AK293091.1 | - | 2 | 979 | ||
AL833127.1 | - | 2 | 417 | |||
AY698056.1 | - | 2 | 417 | |||
AY775731.1 | - | 1 | 525 | |||
BC022401.1 | - | 2 | 472 | 4694856 |