Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKAR SIRNA Assays ›
Gene Symbol:
ANKAR
Gene Name:
ankyrin and armadillo repeat containing
Gene Aliases:
-
Chromosome Location:
Chr.2: 189674292 - 189763059 on Build GRCh38
UniGene ID:
Hs.60772
Species:
Human
siRNA ID
HSS175639
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKAR
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_144708.3
NP_653309.3
3
782
XM_011510673.2
XP_011508975.1
3
1268
XM_011510674.1
XP_011508976.1
3
1268
XM_011510675.2
XP_011508977.1
3
1268
XM_011510676.2
XP_011508978.1
3
1268
XM_011510677.2
XP_011508979.1
3
1268
XM_011510679.2
XP_011508981.1
3
1268
XM_011510680.1
XP_011508982.1
3
1268
XM_011510681.1
XP_011508983.1
3
1268
XM_011510682.2
XP_011508984.1
3
1268
XM_011510685.2
XP_011508987.1
3
824
XM_011510686.2
XP_011508988.1
3
1268
XM_011510687.2
XP_011508989.1
2
632
XM_011510688.1
XP_011508990.1
3
1268
XM_011510689.2
XP_011508991.1
3
1268
XM_011510692.2
XP_011508994.1
2
632
XM_011510693.2
XP_011508995.1
2
632
XM_011510694.2
XP_011508996.1
2
632
XM_017003413.1
XP_016858902.1
3
1268
XM_017003414.1
XP_016858903.1
3
1268
XM_017003415.1
XP_016858904.1
2
632
XM_017003416.1
XP_016858905.1
3
824
XM_017003417.1
XP_016858906.1
3
1268
XM_017003418.1
XP_016858907.1
2
632
XM_017003419.1
XP_016858908.1
2
632
XM_017003420.1
XP_016858909.1
2
632
GenBank mRNA
AJ549812.1
-
2
481
AK098551.1
-
2
247
AK126073.1
-
3
782
AK307328.1
-
2
249
BC037215.1
-
2
431
BC044907.1
-
2
634
4836761
BC047413.1
-
2
172
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-zl9nd:80/10.239.167.245:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0