Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033045.3 | NP_001028217.1 | 3 | 640 | ||
NM_001267050.1 | NP_001253979.1 | 3 | 537 | |||
NM_001267051.1 | NP_001253980.1 | 2 | 491 | |||
NM_152529.6 | NP_689742.4 | 2 | 491 | |||
XM_017003485.1 | XP_016858974.1 | 4 | 682 | |||
XM_017003486.1 | XP_016858975.1 | 2 | 543 | |||
XM_017003487.1 | XP_016858976.1 | 3 | 544 | |||
GenBank mRNA | AK091200.1 | - | 2 | 432 | ||
AK307896.1 | - | 2 | 483 | |||
AK315125.1 | - | 2 | 432 | |||
AL834154.1 | - | 3 | 589 | |||
BC028730.1 | - | 3 | 586 | 4813561 | ||
BC035037.1 | - | 3 | 548 | 5260361 | ||
BC036681.1 | - | 2 | 437 | |||
BX537947.1 | - | 2 | 484 |