Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DMTN SIRNA Assays ›
Gene Symbol:
DMTN
Gene Name:
dematin actin binding protein
Gene Aliases:
DMT, EPB49
Chromosome Location:
Chr.8: 22048955 - 22082527 on Build GRCh38
UniGene ID:
Hs.106124
Species:
Human
siRNA ID
HSS176581
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DMTN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001114135.4
NP_001107607.1
15, 16
1388
NM_001114136.2
NP_001107608.1
15, 16
1344
NM_001114137.3
NP_001107609.1
14, 15
1452
NM_001114138.2
NP_001107610.1
14, 15
1439
NM_001114139.3
NP_001107611.1
13, 14
1336
NM_001302816.2
NP_001289745.1
14, 15
1572
NM_001302817.2
NP_001289746.1
13, 14
1357
NM_001323378.1
NP_001310307.1
15, 16
1477
NM_001323380.1
NP_001310309.1
15, 16
1384
NM_001323381.1
NP_001310310.1
14, 15
1493
NM_001323382.1
NP_001310311.1
15, 16
1423
NM_001323383.1
NP_001310312.1
14, 15
1278
NM_001323384.1
NP_001310313.1
14, 15
1411
NM_001323385.1
NP_001310314.1
13, 14
1427
NM_001323387.1
NP_001310316.1
13, 14
1376
NM_001323388.1
NP_001310317.1
13, 14
1224
NM_001323389.1
NP_001310318.1
14, 15
1432
NM_001323390.1
NP_001310319.1
14, 15
1299
NM_001323391.1
NP_001310320.1
14, 15
1402
NM_001323392.1
NP_001310321.1
14, 15
1563
NM_001323393.1
NP_001310322.1
14, 15
1269
NM_001323394.1
NP_001310323.1
14, 15
1430
NM_001323395.1
NP_001310324.1
14, 15
1309
NM_001323396.1
NP_001310325.1
14, 15
1313
NM_001323397.1
NP_001310326.1
14, 15
1481
NM_001323398.1
NP_001310327.1
12, 13
1291
NM_001323399.1
NP_001310328.1
12, 13
1158
NM_001323400.1
NP_001310329.1
11, 12
1307
NM_001323401.1
NP_001310330.1
12, 13
1331
NM_001978.4
NP_001969.2
15, 16
1638
XM_005273432.1
XP_005273489.1
15, 16
1370
XM_005273433.1
XP_005273490.1
15, 16
1302
XM_005273442.1
XP_005273499.1
14, 15
1154
XM_011544435.1
XP_011542737.1
14, 15
1563
XM_017013189.1
XP_016868678.1
14, 15
1245
XM_017013190.1
XP_016868679.1
13, 14
1497
XM_017013191.1
XP_016868680.1
13, 14
1251
XM_017013192.1
XP_016868681.1
13, 14
1132
XM_017013193.1
XP_016868682.1
13, 14
1330
XM_017013194.1
XP_016868683.1
13, 14
1197
XM_017013195.1
XP_016868684.1
13, 14
1497
XM_017013196.1
XP_016868685.1
13, 14
1100
XM_017013197.1
XP_016868686.1
13, 14
1331
XM_017013198.1
XP_016868687.1
13, 14
1173
GenBank mRNA
AB209718.1
-
11, 12
864
AK055842.1
-
14, 15
1236
AK091581.1
-
13, 14
1251
AK289650.1
-
15, 16
1270
AK295452.1
-
13, 14
1243
BC006318.2
-
15, 16
1387
4125804
BC017445.1
-
15, 16
1387
3162732
BC052805.1
-
14, 15
1362
6269658
BT007396.1
-
14, 15
1115
L19713.1
-
14, 15
1499
U28389.1
-
15, 16
1565
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
cytoskeleton organization
regulation of cell shape
regulation of lamellipodium assembly
positive regulation of fibroblast migration
negative regulation of peptidyl-threonine phosphorylation
negative regulation of cell-substrate adhesion
actin cytoskeleton organization
positive regulation of blood coagulation
regulation of actin cytoskeleton organization
negative regulation of peptidyl-serine phosphorylation
calcium-mediated signaling using intracellular calcium source
calcium-mediated signaling using extracellular calcium source
erythrocyte development
negative regulation of peptidyl-tyrosine phosphorylation
actin filament bundle assembly
regulation of filopodium assembly
actin filament capping
negative regulation of focal adhesion assembly
transmembrane transport
protein secretion by platelet
cellular response to calcium ion
cellular response to cAMP
positive regulation of wound healing
negative regulation of protein targeting to membrane
actin filament reorganization
negative regulation of substrate adhesion-dependent cell spreading
positive regulation of substrate adhesion-dependent cell spreading
positive regulation of platelet aggregation
positive regulation of integrin-mediated signaling pathway
actin binding
receptor binding
protein binding
spectrin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-2g8tf:80/10.239.165.190:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0