Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145306.2 | NP_660349.1 | 4 | 494 | ||
XM_005269606.1 | XP_005269663.1 | 4 | 778 | |||
XM_005269608.3 | XP_005269665.1 | 3 | 432 | |||
XM_011539455.2 | XP_011537757.1 | 4 | 1035 | |||
XM_017015836.1 | XP_016871325.1 | 5 | 666 | |||
GenBank mRNA | BC013587.1 | - | 4 | 483 | 3896656 | |
CK903062.1 | - | 3 | 433 |