Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014909.4 | NP_055724.1 | 2, 3 | 1320 | ||
XM_017021087.1 | XP_016876576.1 | 4, 5 | 395 | |||
XM_017021088.1 | XP_016876577.1 | 3, 4 | 352 | |||
XM_017021089.1 | XP_016876578.1 | 3, 4 | 211 | |||
XM_017021090.1 | XP_016876579.1 | 2, 3 | 144 | |||
XM_017021091.1 | XP_016876580.1 | 2, 3 | 316 | |||
GenBank mRNA | AK291377.1 | - | 2, 3 | 470 | ||
BC051896.1 | - | 2, 3 | 614 | 5170122 |