Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QPRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318249.1 | NP_001305178.1 | 2 | 230 | ||
NM_001318250.1 | NP_001305179.1 | 3 | 322 | |||
NM_014298.4 | NP_055113.2 | 3 | 766 | |||
GenBank mRNA | AK090801.1 | - | 1 | 1230 | ||
AK293195.1 | - | 2 | 230 | |||
BC005060.1 | - | 3 | 603 | 2960170 | ||
BC010033.2 | - | 3 | 595 | 3506460 | ||
BC018910.2 | - | 3 | 620 | 3959973 | ||
BP314322.1 | - | 3 | 148 | |||
BT007231.1 | - | 2 | 576 | |||
D78177.1 | - | 2 | 576 | |||
EU794638.1 | - | 3 | 595 | |||
KJ534923.1 | - | 2 | 576 |