Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK8IP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012324.4 | NP_036456.1 | 5 | 1865 | ||
XM_011530679.2 | XP_011528981.1 | 5 | 1757 | |||
XM_011530680.2 | XP_011528982.1 | 5 | 1643 | |||
XM_011530681.2 | XP_011528983.1 | 5 | 1622 | |||
GenBank mRNA | AF136382.1 | - | 5 | 1848 | ||
AF218778.1 | - | 3 | 1650 | |||
AK294265.1 | - | 6 | 924 | |||
AL021708.1 | - | 1 | 868 | |||
BC009940.2 | - | 4 | 615 | 4100637 | ||
BC047527.1 | - | 5 | 1778 | 5277237 |