Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000149.3 | NP_000140.1 | 3 | 698 | ||
NM_001097639.1 | NP_001091108.1 | 3 | 350 | |||
NM_001097640.1 | NP_001091109.1 | 3 | 296 | |||
NM_001097641.1 | NP_001091110.1 | 2 | 157 | |||
XM_011527865.1 | XP_011526167.1 | 3 | 676 | |||
XM_011527866.1 | XP_011526168.1 | 3 | 337 | |||
XM_011527867.1 | XP_011526169.1 | 3 | 306 | |||
GenBank mRNA | AF131913.1 | - | 2 | 229 | ||
AK291852.1 | - | 3 | 325 | |||
BC074836.2 | - | 2 | 102 | 30915328 | ||
BC074837.2 | - | 2 | 102 | 30915548 | ||
BC108675.1 | - | 2 | 154 | 5187745 | ||
DA876997.1 | - | 3 | 271 | |||
DA930897.1 | - | 3 | 350 | |||
U27326.1 | - | 3 | 289 | |||
U27327.1 | - | 3 | 343 | |||
U27328.1 | - | 4 | 692 | |||
X53578.1 | - | 2 | 132 |