Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320466.1 | NP_001307395.1 | 2, 3 | 562 | ||
NM_001320467.1 | NP_001307396.1 | 2, 3 | 454 | |||
NM_001320468.1 | NP_001307397.1 | 2, 3 | 562 | |||
NM_173570.4 | NP_775841.2 | 2, 3 | 454 | |||
NR_135271.1 | 2, 3 | 562 | ||||
XM_005247269.1 | XP_005247326.1 | 2, 3 | 398 | |||
XM_006713562.3 | XP_006713625.1 | 2, 3 | 398 | |||
XM_011512618.2 | XP_011510920.1 | 2, 3 | 402 | |||
XM_011512619.1 | XP_011510921.1 | 2, 3 | 398 | |||
XM_017006084.1 | XP_016861573.1 | 2, 3 | 378 | |||
XM_017006085.1 | XP_016861574.1 | 2, 3 | 377 | |||
XM_017006086.1 | XP_016861575.1 | 2, 3 | 398 | |||
XM_017006087.1 | XP_016861576.1 | 2, 3 | 486 | |||
GenBank mRNA | AK127025.1 | - | 2, 3 | 398 | ||
AK127996.1 | - | 3, 4 | 399 | |||
BC035230.1 | - | 2, 3 | 454 | 4824088 |