Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G4F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213600.3 | NP_998765.3 | 3 | 346 | ||
NR_033151.1 | 2 | 273 | ||||
GenBank mRNA | AK122623.1 | - | 3 | 317 | ||
AK172836.1 | - | 3 | 346 | |||
AK304368.1 | - | 3 | 328 | |||
BC035335.1 | - | 2 | 243 | |||
BC131531.1 | - | 3 | 323 | 40036909 | ||
BC142682.1 | - | 3 | 317 | 40148418 | ||
BC146648.1 | - | 3 | 317 | 40147152 |