Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199809.1 | NP_001186738.1 | 5 | 648 | ||
NM_001199811.1 | NP_001186740.1 | 6 | 795 | |||
NM_001199812.1 | NP_001186741.1 | 6 | 795 | |||
NM_015434.3 | NP_056249.1 | 6 | 795 | |||
NR_037667.1 | 7 | 829 | ||||
XM_011509396.2 | XP_011507698.1 | 6 | 680 | |||
XM_017000962.1 | XP_016856451.1 | 6 | 680 | |||
GenBank mRNA | AK001363.1 | - | 6 | 655 | ||
AK001598.1 | - | 6 | 677 | |||
AK022509.1 | - | 6 | 655 | |||
AK297225.1 | - | 5 | 527 | |||
BC020523.2 | - | 6 | 671 | 3917030 | ||
BC030716.1 | - | 6 | 650 | 3909231 | ||
BC033918.1 | - | 6 | 654 | 3920829 |