Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015457.2 | NP_056272.2 | 8 | 2095 | ||
XM_011544899.1 | XP_011543201.1 | 8 | 2025 | |||
XM_011544900.1 | XP_011543202.1 | 8 | 1933 | |||
XM_011544901.1 | XP_011543203.1 | 8 | 2056 | |||
XM_017017498.1 | XP_016872987.1 | 7 | 2139 | |||
GenBank mRNA | AB051535.1 | - | 8 | 2087 | ||
AK023130.1 | - | 7 | 1252 | |||
AK172807.1 | - | 4 | 450 | |||
AY894889.1 | - | 8 | 2201 | |||
BC026967.1 | - | 7 | 838 | 3913317 | ||
BX647707.1 | - | 6 | 1087 |