Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153615.1 | NP_705843.1 | 3 | 1770 | ||
GenBank mRNA | AK131340.1 | - | 5 | 2208 | ||
AY101396.1 | - | 3 | 1770 | |||
BC101108.3 | - | 2 | 1450 | 40011557 | ||
BC101109.2 | - | 2 | 1450 | 40011564 | ||
BC101110.3 | - | 3 | 1325 |
Gene Symbol: GUSBP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125808.1 | - | 1 | 254 | ||
CR933599.1 | - | 1 | 287 |