Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP3M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320263.1 | NP_001307192.1 | 10 | 1686 | ||
NM_001320264.1 | NP_001307193.1 | 8 | 1525 | |||
NM_001320265.1 | NP_001307194.1 | 8 | 1237 | |||
NM_012095.5 | NP_036227.1 | 8 | 1399 | |||
NM_207012.3 | NP_996895.1 | 9 | 1525 | |||
NR_135191.1 | 8 | 1421 | ||||
GenBank mRNA | AF092092.1 | - | 7 | 1065 | ||
AK026983.1 | - | 8 | 1134 | |||
AK299352.1 | - | 8 | 988 | |||
AK315597.1 | - | 8 | 1107 | |||
AL833820.1 | - | 6 | 704 | |||
BC026232.1 | - | 9 | 1284 | 4807259 | ||
BC067127.1 | - | 8 | 1495 | 6169822 | ||
DA174180.1 | - | 2 | 215 |