Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256182.1 | NP_001243111.1 | 10 | 4413 | ||
NM_001256183.1 | NP_001243112.1 | 9 | 4339 | |||
NM_013275.5 | NP_037407.4 | 9 | 4342 | |||
XM_006721181.1 | XP_006721244.1 | 9 | 4119 | |||
XM_006721184.2 | XP_006721247.1 | 8 | 4157 | |||
XM_011523051.2 | XP_011521353.1 | 10 | 4402 | |||
XM_011523053.2 | XP_011521355.1 | 10 | 4508 | |||
XM_011523054.2 | XP_011521356.1 | 10 | 4422 | |||
XM_011523055.2 | XP_011521357.1 | 11 | 4707 | |||
XM_011523056.2 | XP_011521358.1 | 10 | 4444 | |||
XM_011523057.2 | XP_011521359.1 | 9 | 4325 | |||
XM_017023182.1 | XP_016878671.1 | 9 | 4775 | |||
XM_017023183.1 | XP_016878672.1 | 10 | 4511 | |||
XM_017023184.1 | XP_016878673.1 | 10 | 4508 | |||
XM_017023185.1 | XP_016878674.1 | 8 | 4149 | |||
XM_017023186.1 | XP_016878675.1 | 12 | 4885 | |||
XM_017023187.1 | XP_016878676.1 | 8 | 4237 | |||
GenBank mRNA | AF130076.1 | - | 1 | 629 | ||
AK057803.1 | - | 1 | 1385 | |||
AY373756.1 | - | 9 | 4345 | |||
AY533563.1 | - | 10 | 4170 | |||
BC058001.1 | - | 1 | 861 |