Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC40A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014585.5 | NP_055400.1 | 7 | 1624 | ||
XM_005246505.1 | XP_005246562.1 | 8 | 1759 | |||
XM_017003938.1 | XP_016859427.1 | 8 | 1873 | |||
GenBank mRNA | AF226614.1 | - | 7 | 1577 | ||
AF231121.1 | - | 7 | 1577 | |||
AK002038.1 | - | 2 | 559 | |||
AK223236.1 | - | 7 | 1599 | |||
AK292146.1 | - | 7 | 1599 | |||
AK314827.1 | - | 7 | 1599 | |||
AL136944.1 | - | 7 | 1587 | |||
BC035893.1 | - | 7 | 1588 | 5213437 | ||
BC037733.1 | - | 7 | 1703 | 4823308 | ||
CR533564.1 | - | 7 | 1273 |