Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005167.5 | NP_005158.5 | 4, 5 | 879 | ||
GenBank mRNA | AB451327.1 | - | 3, 4 | 234 | ||
AB451471.1 | - | 3, 4 | 234 | |||
AK093270.1 | - | 4, 5 | 859 | |||
AL133644.1 | - | 5, 6 | 1643 | |||
BC011803.2 | - | 4, 5 | 803 | |||
BC064628.1 | - | 4, 5 | 809 | |||
BC073828.1 | - | 4, 5 | 871 | 5244363 |