Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286509.1 | NP_001273438.1 | 4 | 755 | ||
NM_001286510.1 | NP_001273439.1 | 4 | 754 | |||
NM_001286511.1 | NP_001273440.1 | 4 | 709 | |||
NM_001286512.1 | NP_001273441.1 | 4 | 758 | |||
NM_001286513.1 | NP_001273442.1 | 3 | 604 | |||
NM_001286517.1 | NP_001273446.1 | 3 | 565 | |||
NM_001286519.1 | NP_001273448.1 | 2 | 410 | |||
NM_001286520.1 | NP_001273449.1 | 4 | 763 | |||
NM_178148.3 | NP_835361.1 | 4 | 759 | |||
GenBank mRNA | AB097021.1 | - | 4 | 716 | ||
AB097039.1 | - | 4 | 716 | |||
AB106538.1 | - | 4 | 607 | |||
AK075456.1 | - | 4 | 688 | |||
AK123749.1 | - | 4 | 656 | |||
AK223028.1 | - | 3 | 409 | |||
AK293251.1 | - | 2 | 377 | |||
AK293344.1 | - | 3 | 561 | |||
AK307572.1 | - | 4 | 694 | |||
AY491520.1 | - | 4 | 725 | |||
BC024288.1 | - | 4 | 723 | 4890949 | ||
BI769501.1 | - | 3 | 495 | |||
CN370928.1 | - | 4 | 655 |