Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198887.2 | NP_942590.1 | 5 | 677 | ||
NR_104456.1 | 5 | 677 | ||||
XM_005266960.4 | XP_005267017.1 | 5 | 627 | |||
XM_005266961.3 | XP_005267018.1 | 5 | 627 | |||
XM_005266962.3 | XP_005267019.1 | 5 | 627 | |||
XM_011535798.2 | XP_011534100.1 | 5 | 627 | |||
XM_011535799.2 | XP_011534101.1 | 5 | 627 | |||
GenBank mRNA | AF514997.1 | - | 5 | 610 | ||
AK074311.1 | - | 632 | ||||
AK095994.1 | - | 5 | 633 | |||
AK304505.1 | - | 4 | 484 | |||
BC040121.1 | - | 3 | 322 | |||
BC065028.1 | - | 5 | 603 | 6042215 | ||
BX647097.1 | - | 5 | 627 |