Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000588.3 | NP_000579.2 | 3 | 332 | ||
GenBank mRNA | BC066272.1 | - | 3 | 325 | 6971773 | |
BC066273.1 | - | 3 | 325 | 6971774 | ||
BC066274.1 | - | 3 | 325 | 6971775 | ||
BC066275.1 | - | 3 | 325 | 6971776 | ||
BC066276.1 | - | 3 | 325 | 6971777 | ||
BC069472.1 | - | 3 | 328 | 7262122 | ||
M14743.1 | - | 3 | 288 | |||
M17115.1 | - | 3 | 317 | |||
M20137.1 | - | 3 | 332 |