Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145026.1 | NP_001138498.1 | 12 | 1846 | ||
XM_017019273.1 | XP_016874762.1 | 16 | 2648 | |||
XM_017019274.1 | XP_016874763.1 | 16 | 2649 | |||
XM_017019275.1 | XP_016874764.1 | 16 | 2649 | |||
GenBank mRNA | DV080607.1 | - | 2 | 195 |
Set Membership: |
G-protein coupled receptor |