Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099666.1 | NP_001093136.1 | 8 | 1207 | ||
XM_005251976.4 | XP_005252033.1 | 7 | 1378 | |||
XM_005251977.4 | XP_005252034.1 | 8 | 1268 | |||
XM_011518639.1 | XP_011516941.1 | 7 | 1252 | |||
XM_011518640.1 | XP_011516942.1 | 8 | 1428 | |||
GenBank mRNA | AK127511.1 | - | 2 | 542 | ||
AL832683.1 | - | 5 | 1032 | |||
BC053622.1 | - | 8 | 1189 |