Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WASH3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097269.1 | - | 1, 2 | 972 | ||
AK300161.1 | - | 3 | 358 | |||
BC063555.1 | - | 1 | 277 |
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011517663.2 | XP_011515965.1 | 1 | 1507 | ||
XM_011517664.2 | XP_011515966.1 | 1 | 1507 | |||
XM_011517665.2 | XP_011515967.1 | 1 | 43 | |||
XM_017014169.1 | XP_016869658.1 | 2 | 271 | |||
XM_017014170.1 | XP_016869659.1 | 2 | 337 |
Gene Symbol: WASH2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY217347.1 | - | 1 | 802 | ||
BC082240.1 | - | 1 | 294 |
Gene Symbol: WASH7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK294377.1 | - | 1 | 53 | ||
AK296182.1 | - | 3 | 293 | |||
AK308540.1 | - | 3 | 358 | |||
AK316487.1 | - | 1 | 346 | |||
BC094698.1 | - | 1 | 149 |